Ethyl-trans-4-oxo-2-butenoate 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 96%
CAS No.: 2960-66-9
Molecular Formula: C6H8O3
Formula Weight: 128.13
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.4550
Melting Point: -9°
Boiling Point: 184-188°
Beilstein Reference: 8762336
UN No.: UN2922
MDL No.: MFCD00236170
Formel:
C₆H₈O₃ Siedepunkt: 184…188 °C Schmelzpunkt: –9 °C Dichte: 1,063 Flammpunkt: 68 °C (154 °F) |
MDL:
MFCD00236170 CAS-Nummer: 2960-66-9 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...