Ethyl (R)-4-chloro-3-hydroxybutyrate 97%, ee 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(R)-Ethyl 4-chloro-3-hydroxybutanoate, (R)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester
L18553.03
L18553.06
L18553.03EA
56.2
EUR
L18553.03
L18553.06
Ethyl (R)-4-chloro-3-hydroxybutyrate 97%, ee 96%
Ethyl (R)-4-chloro-3-hydroxybutyrate
Purity: 97%
CAS No.: 90866-33-4
Molecular Formula: C6H11ClO3
Formula Weight: 166.61
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₆H₁₁ClO₃ Molecular Weight: 166,60 g/mol Siedepunkt: 93…95 °C (5 mmHg) Dichte: 1,190 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00211242 CAS-Nummer: 90866-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...