D-Epoxone 98%

Lieferant: Thermo Scientific

Synonyme: 1,2:4,5-Di-O-Isopropylidene-Beta-D-Erythro-2,3-Hexodiulo-2,6-Pyranose

L18989.03 L18989.06
L18989.03EA 49.7 EUR
L18989.03 L18989.06
D-Epoxone 98%
D-Epoxone

Purity: 98% CAS No.: 18422-53-2 Molecular Formula: C12H18O6 Formula Weight: 258.27 Melting Point: 101-103° Beilstein Reference: 1319073 MDL No.: MFCD00063383

Formel: C₁₂H₁₈O₆
Molecular Weight: 258,27 g/mol
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063383
CAS-Nummer: 18422-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR