Methyl-3-quinuclidinecarboxylate hydrochloride ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L19132.03 L19132.MD
L19132.03EA 184 EUR
L19132.03 L19132.MD
Methyl-3-quinuclidinecarboxylate hydrochloride ≥98%
Methyl-3-quinuclidinecarboxylate hydrochloride

Purity: 98+% CAS No.: 54954-73-3 Molecular Formula: C9H15NO2.HCl Formula Weight: 205.69 Melting Point: ca 160° dec. Beilstein Reference: 4272456 MDL No.: MFCD01073582

Formel: C₉H₁₅NO₂·HCl
Molecular Weight: 205.69 g/mol
Schmelzpunkt: ∼ 160 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01073582
CAS-Nummer: 54954-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR