2-(4-Benzyloxyphenyl)ethanol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-(4-(Benzyloxy)phenyl)ethanol
L19217.14
L19217.06
L19217.14EA
193
EUR
L19217.14
L19217.06
2-(4-Benzyloxyphenyl)ethanol ≥98%
2-(4-Benzyloxyphenyl)ethanol
Purity: 98+%
CAS No.: 61439-59-6
Molecular Formula: C15H16O2
Formula Weight: 228.29
Melting Point: 85-87°
Beilstein Reference: 2054624
MDL No.: MFCD00017532
Formel:
C₁₅H₁₆O₂ Molecular Weight: 228.29 g/mol Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017532 CAS-Nummer: 61439-59-6 EINECS: 262-795-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...