Methyl-L-prolinathydrochlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
H-Pro-OMe.HCl, H-Pro-OMe·HCl
L19290.03
L19290.06
L19290.03EA
29.3
EUR
L19290.03
L19290.06
Methyl-L-prolinathydrochlorid ≥98%
L-Proline methyl ester hydrochloride
Purity: 98+%
CAS No.: 2133-40-6
Molecular Formula: C6H11NO2.HCl
Formula Weight: 165.62
Storage Temperature: 2°C to 8°C
Melting Point: 71-78°
Beilstein Reference: 3596045
Optical Rotation: -33° (c=1 in water)
MDL No.: MFCD00012708
Formel:
C₆H₁₁NO₂·HCl Molecular Weight: 165,6 g/mol Schmelzpunkt: 71…78 °C |
MDL:
MFCD00012708 CAS-Nummer: 2133-40-6 EINECS: 218-363-0 |