(S)-(+)-2-Amino-1-phenylethanol 97%
Lieferant: Thermo Scientific
|
Gefahr
|
L19404.03
L19404.06
L19404.03EA
72.5
EUR
L19404.03
L19404.06
(S)-(+)-2-Amino-1-phenylethanol 97%
(S)-(+)-2-Amino-1-phenylethanol
Purity: 97%
CAS No.: 56613-81-1
Molecular Formula: C8H11NO
Formula Weight: 137.18
Melting Point: 57-63°
Boiling Point: 160°/17mm
Beilstein Reference: 2412760
Optical Rotation: +42° (c=2 in ethanol)
UN No.: UN3259
MDL No.: MFCD00239405
Formel:
C₈H₁₁NO Molecular Weight: 137,182 g/mol Siedepunkt: 160 °C (17 mmHg) Schmelzpunkt: 57…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00239405 CAS-Nummer: 56613-81-1 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...