1-Benzyl-3-pyrroline 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Benzyl-2,5-dihydropyrrole
L19496.03
L19496.06
L19496.03EA
32.5
EUR
L19496.03
L19496.06
1-Benzyl-3-pyrroline 98%
1-Benzyl-3-pyrroline
Purity: 98+%
CAS No.: 6913-92-4
Molecular Formula: C11H13N
Formula Weight: 159.23
Storage Temperature: 2°C to 8°C
Flash Point: 92°(197°F)
Refractive Index: 1.5420
Boiling Point: 60°/1mm
Beilstein Reference: 111101
UN No.: UN2735
MDL No.: MFCD00012324
Formel:
C₁₁H₁₃N Molecular Weight: 159,23 g/mol Siedepunkt: 60 °C (1 mmHg) Dichte: 0,837 Flammpunkt: 92 °C (197 °F) |
MDL:
MFCD00012324 CAS-Nummer: 6913-92-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...