3-Benzo[b]furylacetonitrile 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Benzofuran-3-ylacetonitrile

L19550.03 L19550.MD
L19550.03EA 132 EUR
L19550.03 L19550.MD
3-Benzo[b]furylacetonitrile 99%
3-Benzo[b]furylacetonitrile

Purity: 99% CAS No.: 52407-43-9 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 38-39° Boiling Point: 120-122°/4mm Beilstein Reference: 1282785 MDL No.: MFCD02183555

Formel: C₁₀H₇NO
Molecular Weight: 157.17 g/mol
Siedepunkt: 120…122 °C (4 mmHg)
Schmelzpunkt: 38…39 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183555
CAS-Nummer: 52407-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR