Benzothiazole-2-carbonyl chloride 95%

Lieferant: Thermo Fisher Scientific

L19631.03 L19631.MD
L19631.03EA 137 EUR
L19631.03 L19631.MD
Benzothiazole-2-carbonyl chloride 95%
Benzothiazole-2-carbonyl chloride

Purity: 95% CAS No.: 67748-61-2 Molecular Formula: C8H4ClNOS Formula Weight: 197.64 Melting Point: 113-115° Beilstein Reference: 130990 UN No.: UN3261 MDL No.: MFCD03659697

Formel: C₈H₄ClNOS
Molecular Weight: 197.64 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03659697
CAS-Nummer: 67748-61-2
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR