(R)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-(R)-PPFA

L19685.04 L19685.MC L19685.ME
L19685.04EA 742 EUR
L19685.04 L19685.MC L19685.ME
(R)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥97%
(R)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine

Purity: 97% CAS No.: 55700-44-2 Molecular Formula: C26H28FeNP Formula Weight: 441.34 Melting Point: 141-143° Optical Rotation: -355° (c=0.6 in ethanol) MDL No.: MFCD00075286

Formel: C₂₆H₂₈FeNP
Molecular Weight: 441,34 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075286
CAS-Nummer: 55700-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR