4-Methyl-β-styrylboronic acid pinacol ester 98%

Lieferant: Thermo Scientific

Synonyme: (E)-4,4,5,5-Tetramethyl-2-(4-methylstyryl)-1,3,2-dioxaborolane

L19698.03 L19698.MD
L19698.03EA 130 EUR
L19698.03 L19698.MD
4-Methyl-β-styrylboronic acid pinacol ester 98%
4-Methyl-beta-styrylboronic acid pinacol ester

Purity: 98% CAS No.: 149777-84-4 Molecular Formula: C15H21BO2 Formula Weight: 244.14 Melting Point: 56-58° Beilstein Reference: 7204616 MDL No.: MFCD03788749

Formel: C₁₅H₂₁BO₂
Molecular Weight: 244,14 g/mol
Schmelzpunkt: 56…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788749
CAS-Nummer: 149777-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR