Methyl-Β-L-arabinopyranoside ≥98%

Lieferant: Thermo Scientific

Synonyme: (2S,3R,4S,5S)-2-Methoxytetrahydro-2H-pyran-3,4,5-triol

L19746.03 L19746.06
L19746.03EA 80.7 EUR
L19746.03 L19746.06
Methyl-Β-L-arabinopyranoside ≥98%
Methyl-Β-L-arabinopyranoside

Purity: 98+% CAS No.: 1825-00-9 Molecular Formula: C6H12O5 Formula Weight: 164.16 Melting Point: 167-172° Beilstein Reference: 4658388 Optical Rotation: +243° (c=2 in water) MDL No.: MFCD00047547

Formel: C₆H₁₂O₅
Molecular Weight: 164,17 g/mol
Schmelzpunkt: 167…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047547
CAS-Nummer: 1825-00-9
EINECS: 217-362-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR