Isoprene monoxide 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-2-vinyloxirane
L19768.06
L19768.14
L19768.06EA
96.6
EUR
L19768.06
L19768.14
Isoprene monoxide 97%
Isoprene monoxide
Purity: 97%
CAS No.: 1838-94-4
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: -8°(17°F)
Refractive Index: 1.4160
Odour: Recognizable
Boiling Point: 79-81°
Beilstein Reference: 104216
UN No.: UN1993
MDL No.: MFCD00075225
Formel:
C₅H₈O Molecular Weight: 84,12 g/mol Siedepunkt: 79…81 °C Dichte: 0,857 Flammpunkt: –8 °C (17 °F) |
MDL:
MFCD00075225 CAS-Nummer: 1838-94-4 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...