Ethyl-(3R)-4-cyan-3-hydroxybutanoat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-4-Cyano-3-hydroxybutyric acid ethyl ester, BUTANOIC ACID, 4-CYANO-3-HYDROXY-, ETHYL ESTER, (3R)-, Ethyl (R)-4-cyano-3-hydroxybutanoate, (R)-ethyl 4-cyano-3-hydroxybutanoate
L19771.06
L19771.14
L19771.06EA
58.3
EUR
L19771.06
L19771.14
Ethyl-(3R)-4-cyan-3-hydroxybutanoat 98%
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate
Purity: 98%
CAS No.: 141942-85-0
Molecular Formula: C7H11NO3
Formula Weight: 157.17
Flash Point:
Formel:
C₇H₁₁NO₃ Molecular Weight: 157,169 g/mol Siedepunkt: 270 °C Dichte: 1,114 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270839 CAS-Nummer: 141942-85-0 EINECS: 430-220-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...