3-Fluoropiperidine hydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L19776.03
L19776.MD
L19776.03EA
348
EUR
L19776.03
L19776.MD
3-Fluoropiperidine hydrochloride 97%
3-Fluoropiperidine hydrochloride
Purity: 97%
CAS No.: 737000-77-0
Molecular Formula: C5H10FN.HCl
Formula Weight: 139.60
Melting Point: ca 227°
Beilstein Reference: 6145604
MDL No.: MFCD03452799
Formel:
C₅H₁₀FN·HCl Molecular Weight: 139.60 g/mol Schmelzpunkt: ∼ 227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03452799 CAS-Nummer: 737000-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...