2-Aminopyridine-4-methanol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol
L19846.03
L19846.06
L19846.03EA
51.7
EUR
L19846.03
L19846.06
2-Aminopyridine-4-methanol 97%
2-Aminopyridine-4-methanol
Purity: 97%
CAS No.: 105250-17-7
Molecular Formula: C6H8N2O
Formula Weight: 124.14
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Cream to pale yellow
Melting Point: 81-85°
Beilstein Reference: 8760550
MDL No.: MFCD03791261
Formel:
C₆H₈N₂O Molecular Weight: 124,143 g/mol Schmelzpunkt: 81…85 °C |
MDL:
MFCD03791261 CAS-Nummer: 105250-17-7 |