(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

L20131.03 L20131.06
L20131.03EA 97.2 EUR
L20131.03 L20131.06
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 97%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

Purity: 97% CAS No.: 113451-59-5 Molecular Formula: C10H18N2O2 Formula Weight: 198.26 Storage Temperature: 2°C to 8°C Melting Point: 74-76° Beilstein Reference: 6460311 Optical Rotation: -44° (c=1 in chloroform) MDL No.: MFCD01569250

Formel: C₁₀H₁₈N₂O₂
Molecular Weight: 198.26 g/mol
Schmelzpunkt: 74…76 °C
MDL: MFCD01569250
CAS-Nummer: 113451-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR