4-(Dimethylamino)piperidine 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N,N-Dimethylpiperidin-4-amine

L20176.06 L20176.14
L20176.06EA 102 EUR
L20176.06 L20176.14
4-(Dimethylamino)piperidine 97%
4-(Dimethylamino)piperidine

Purity: 97% CAS No.: 50533-97-6 Molecular Formula: C7H16N2 Formula Weight: 128.22 Refractive Index: 1.4760 Boiling Point: 187° Beilstein Reference: 1190 UN No.: UN2735 MDL No.: MFCD00023144

Formel: C₇H₁₆N₂
Molecular Weight: 128,219 g/mol
Siedepunkt: 187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023144
CAS-Nummer: 50533-97-6
EINECS: 256-617-2
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR