(3,4-Difluorphenyl)(oxo)acetaldehydhydrat 98% (Trockengewicht)

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(3,4-Difluorophenyl)-2-oxoacetaldehyde hydrate

L20271.03 L20271.06
L20271.03EA 104 EUR
L20271.03 L20271.06
(3,4-Difluorphenyl)(oxo)acetaldehydhydrat 98% (Trockengewicht)
3,4-Difluorophenylglyoxal hydrate

Purity: 98% CAS No.: 79784-34-2 Molecular Formula: C8H4F2O2.xH2O Formula Weight: 170.12(anhy) Melting Point: 85-90° Beilstein Reference: 2574671 MDL No.: MFCD03453081

Formel: C8H6F2O3
Molecular Weight: 188,13 g/mol
Schmelzpunkt: 85…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03453081
CAS-Nummer: 79784-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR