4-Fluorophenylglyoxal hydrate, hydrate 98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 2-(4-Fluorophenyl)-2-oxoacetaldehyde hydrate, 4-Fluorophenyl(oxo)glyoxal hydrate
L20272.03 L20272.06
L20272.03EA 77.5 EUR
L20272.03 L20272.06
4-Fluorophenylglyoxal hydrate, hydrate 98%
4-Fluorophenylglyoxal hydrate
Purity: 98% CAS No.: 403-32-7 Molecular Formula: C8H5FO2.xH2O Formula Weight: 152.13(anhy) Melting Point: 84-88° Beilstein Reference: 2354778 MDL No.: MFCD01320770
Formel: C₈H₇FO₃
Molecular Weight: 170.140 g/mol
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320770
CAS-Nummer: 403-32-7