3,4-(Methylenedioxy)phenylglyoxal hydrate 97% (Trockengewicht)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,4-(Methylenedioxy)phenylglyoxal hydrate, 2-(Benzo[d][1,3]dioxol-5-yl)-2-oxoacetaldehyde hydrate
L20273.03
L20273.06
L20273.03EA
73.6
EUR
L20273.03
L20273.06
3,4-(Methylenedioxy)phenylglyoxal hydrate 97% (Trockengewicht)
3,4-(Methylenedioxy)phenylglyoxal hydrate
Purity: 97%
CAS No.: 65709-23-1
Molecular Formula: C9H6O4.xH2O
Formula Weight: 178.15(anhy)
Melting Point: 127-131°
Beilstein Reference: 7703971
MDL No.: MFCD06801031
Formel:
C₉H₆O₄·xH₂O Molecular Weight: 196,16 g/mol Schmelzpunkt: 127…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801031 CAS-Nummer: 65709-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...