4-(Trifluoromethyl)phenylglyoxal hydrate 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde hydrate, Oxo[4-(trifluoromethyl)phenyl]acetaldehyde hydrate (1:1)

L20275.03 L20275.06
L20275.03EA 143 EUR
L20275.03 L20275.06
4-(Trifluoromethyl)phenylglyoxal hydrate 98%
4-(Trifluoromethyl)phenylglyoxal hydrate

Purity: 98% CAS No.: 1736-56-7 Molecular Formula: C9H5F3O2.xH2O Formula Weight: 202.13(anhy) Melting Point: 83-85° Beilstein Reference: 2261131 MDL No.: MFCD05664098

Formel: C₉H₅F₃O₂·xH₂O
Molecular Weight: 202.13 g/mol (anhy)
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664098
CAS-Nummer: 1736-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR