3-Cyan-4-fluorphenylboronsäurepinakolester 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 2-(3-Cyano-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Cyano-4-fluorophenylboronic acid Pinacol ester, 3-Cyano-4-fluorophenylboronic acid, pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

H62245.03 H62245.06
H62245.03EA 164 EUR
H62245.03 H62245.06
3-Cyan-4-fluorphenylboronsäurepinakolester 96%
3-Cyan-4-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247,08 g/mol
Schmelzpunkt: 75…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795681
CAS-Nummer: 775351-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR