Polymethylhydrosiloxan
Lieferant: Thermo Fisher Scientific
Synonyme: Methyl hydrogen polysiloxane, PMHS
L14561.18 L14561.30 L14561.0B
L14561.18EA 25.3 EUR
L14561.18 L14561.30 L14561.0B
Polymethylhydrosiloxan
Polymethylhydrosiloxan, kinematische Viskosität 30 cSt (25°C)
Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.
CAS No.: 63148-57-2 Molecular Formula: (CH3)3SiO[(CH3)HSiO]nSi(CH3) Formula Weight: ca 1900 Flash Point: 204°(399°F) Refractive Index: 1.3980
Formel: (CH₃)₃SiO[(CH₃)HSiO]nSi(CH₃)
Siedepunkt: ∼140 °C (0.003 hPa)
Schmelzpunkt: –55 °C
Dichte: 1.006
Flammpunkt: 204 °C (399 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00084478
CAS-Nummer: 63148-57-2
Bestellen
TECHNISCHE DATEN
Environmentally Preferable