Saccharin Natrium Dihydrat, Pulver USP
Lieferant: Spectrum Chemical
Synonyme: Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide dihydrate, Sodium Saccharin, Dihydrate, Saccharin sodium salt, o-sulphobenzimide sodium salt dihydrate, Saccharin Sodium Salt Dihydrate, o-Benzoic sulphimide sodium salt
SO200-12KG SO200-2.5KG
SPCMSO200-12KGEA 0 EUR
SPCMSO200-12KG SPCMSO200-2.5KG
Saccharin Natrium Dihydrat, Pulver USP
Saccharin Natrium Dihydrat
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel: C₇H₅NNaO₃S·2H₂O
Molecular Weight: 242,21 g/mol
MDL: MFCD00151213
CAS-Nummer: 6155-57-3
EINECS: 204-886-1
Bestellen
Spezifikation Testergebnisse
Assay 99% Min.