Zink-D-gluconat ≥97,0% USP
Lieferant: Spectrum Chemical
Synonyme:
Zinc gluconate
Z1077-12KG
Z1077-50KGBL
Z1077-2.5KG
SPCMZ1077-12KGEA
2640
EUR
SPCMZ1077-12KG
SPCMZ1077-50KGBL
SPCMZ1077-2.5KG
Zink-D-gluconat ≥97,0% USP
Zink-D-gluconat
ZINC GLUCONATE, USP is a zinc salt of gluconic acid can also be manufactured by fermenting glucose. It comes as a white powder and is used in cold treating lozenges and as one of the more effective ways of delivering zinc as a dietary supplement. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₁₂H₂₂O₁₄Zn Molecular Weight: 455,69 g/mol |
MDL:
MFCD00868110 CAS-Nummer: 4468-02-4 EINECS: 224-736-9 |
Spezifikation Testergebnisse
Assay (Anhydrous Basis) | 97.0 - 102.0 % |
pH of a 1 in 100 Solution | 5.5 - 7.5 |
Water | max. 11.6 % |
Chloride (Cl) | max. 0.05 % |
Sulfate | max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...