N-Acetyl-D-galactosamine, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: 2-Acetamido-2-deoxy-D-galactose, D-N-Acetylgalactosamine, 2-Acetamino-2-deoxy-alpha-D-galactopyranose, GalNAc, N-Acetylchondrosamine

02100064.1 0210006425 0210006405 02100064.5 0210006401
ICNA02100064.1EA 74.6 EUR
ICNA02100064.1 ICNA0210006425 ICNA0210006405 ICNA02100064.5 ICNA0210006401
N-Acetyl-D-galactosamine, weißes Pulver
N-Acetyl-D-galactosamine

Soluble in water (100 mg/ml - clear, colourless solution).


N-Acetyl-D-galactosamine(GalNAc), an aminosugar, is a component of many O-linked and N-linked glycan structures.


N-Acetyl-D-galactosamine is the intial O-linked sugar to many serine and threonine residues in protein glycosylations.


Soluble in water (100 mg/ml - clear, colourless solution).

Formel: C₈H₁₅NO₆
Molecular Weight: 221,21 g/mol
Schmelzpunkt: 160…161 °C (Lit.)
MDL: MFCD00065372
CAS-Nummer: 14215-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR