(±)-2-Aminobuttersäure, Pulver

Lieferant: MP Biomedicals

Synonyme: H-DL-Abu-OH, DL-2-Aminobutyric acid, DL-α-Amino-N-Butyric Acid

0210043090 0210043025 0210043080
ICNA0210043090EA 847 EUR
ICNA0210043090 ICNA0210043025 ICNA0210043080
(±)-2-Aminobuttersäure, Pulver
(±)-2-Aminobuttersäure

Soluble in water (5%). Sparingly soluble in alcohol; insoluble in ether.

Formel: C₄H₉NO₂
Molecular Weight: 103,12 g/mol
MDL: MFCD00008093
CAS-Nummer: 2835-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR