Oleoyl coenzyme A dipotassium salt ≥90%, Pulver

Lieferant: MP Biomedicals

Synonyme: cis-9-Octadecenoyl coenzyme A potassium salt

0210090410 0210090450 0210090401 0210090405
ICNA0210090410EA 361 EUR
ICNA0210090410 ICNA0210090450 ICNA0210090401 ICNA0210090405
Oleoyl coenzyme A dipotassium salt ≥90%, Pulver
Oleoyl coenzyme A dipotassium salt

Soluble in water.


Coenzyme A functions as an acyl group carrier, acetyl-CoA. Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.

Formel: C₃₉H₆₆N₇O₁₇P₃SK₂·3H₂O
Molecular Weight: 1162,2 g/mol
MDL: MFCD00871328
CAS-Nummer: 1716-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR