5-Phospho-D-ribose 1-diphosphate pentasodium salt, weißes Pulver
Lieferant: MP Biomedicals
Achtung
Synonyme: 5-Phosphoribosyl 1-pyrophosphate pentasodium salt, α-D-Ribosyl diphosphate 5-phosphate pentasodium salt, 5-Phospho-α-D-ribosyl diphosphate
0210098180 0210098110 0210098125
ICNA0210098180EA 0 EUR
ICNA0210098180 ICNA0210098110 ICNA0210098125
5-Phospho-D-ribose 1-diphosphate pentasodium salt, weißes Pulver
5-Phospho-D-ribose 1-diphosphate pentasodium salt
5-Phosphorylribose-1-pyrophosphate Sodium Salt Hydrate has been used in a study to assess the relationship between the muscarinic receptor cationic current and internal calcium in guinea-pig jejunal smooth muscle cells. It has also been used in a study to investigate inhibitors of the bacterial cell wall biosynthesis of the enzyme Mur D.
Used in a study to assess the relationship between the muscarinic receptor cationic current and internal calcium in guinea-pig jejunal smooth muscle cells.
Soluble in water (50 mg/ml - clear to slightly hazy yellowish solution).
Formel: C₅H₈Na₅O₁₄P₃
Molecular Weight: 499,98 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00084655
CAS-Nummer: 108321-05-7
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Water Content Report result
Sodium Content Report result
Expiration Date Month/Day/Year