Disodium UDP-glucose ≥90%, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: ((2R,3S,4R,5R)-5-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl ((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)diphosphate sodium, UDPG, UDP-α-D-Glucose disodium salt, UDP-Glucose, Uridine 5′-diphosphoglucose disodium salt, Uridine-5'-diphosphoglucose, Uridine-5′-diphosphoglucose disodium salt

0210120880 0210120883 0210120891 0210120894
ICNA0210120880EA 89.9 EUR
ICNA0210120880 ICNA0210120883 ICNA0210120891 ICNA0210120894
Disodium UDP-glucose ≥90%, weißes Pulver
Disodium UDP-glucose

Uridine-5'-diphosphoglucose Disodium Salt is a biosynthetic product produced by the reaction of UTP and glucose-1-phosphate catalysed by uridyl transferase.


  • Loss on Drying: 0,052
  • UV content: 0,75


It is freely soluble in water (50 mg/ml in water): clear, colourless solution; sparingly soluble in methanol; practically insoluble in alcohol, ether, pyridine.


Freely soluble in water; sparingly soluble in methanol; practically insoluble in alcohol, ether, pyridine.


Bitte beachten Research Use Only (RUO).

Formel: C₁₅H₂₂N₂Na₂O₁₇P₂
Molecular Weight: 610.26 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077895
CAS-Nummer: 28053-08-9

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR