Adenosine-5'-diphosphate monopotassium salt dihydrate ≥98%, Pulver

Lieferant: MP Biomedicals

Synonyme: 5'-ADP-K dihydrate, Potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-4-hydroxy-5-(((hydroxy(phosphonooxy)phosphoryl)oxy)methyl)tetrahydrofuran-3-olate dihydrate, ADP-K dihydrate

02150259.5 0215025901 0215025905
ICNA02150259.5EA 53.3 EUR
ICNA02150259.5 ICNA0215025901 ICNA0215025905
Adenosine-5'-diphosphate monopotassium salt dihydrate ≥98%, Pulver
Adenosine-5'-diphosphate monopotassium salt dihydrate

Soluble in water (5 mg/ml - clear, colourless solution).

Formel: C₁₀H₁₄KN₅O₁₀P₂·2H₂O
Molecular Weight: 501,33 g/mol
MDL: MFCD00150944
CAS-Nummer: 72696-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR