L-Aspartic acid magnesium salt, Pulver

Lieferant: MP Biomedicals

0215040691 0215040625
ICNA0215040691EA 476 EUR
ICNA0215040691 ICNA0215040625
L-Aspartic acid magnesium salt, Pulver
L-Aspartic acid magnesium salt

Freely soluble in water.

Formel: C₈H₁₂MgN₂O₈
CAS-Nummer: 18962-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR