Chlorogenic acid, Pulver

Lieferant: MP Biomedicals

Synonyme: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

02150618.1 0215061801 0215061805
ICNA02150618.1EA 36.8 EUR
ICNA02150618.1 ICNA0215061801 ICNA0215061805
Chlorogenic acid, Pulver
Chlorogenic acid

Soluble in water or ethanol (25 mg/ml - clear to hazy, yellow solution).

Formel: C₁₆H₁₈O₉
Molecular Weight: 354,31 g/mol
MDL: MFCD00003862
CAS-Nummer: 327-97-9

Bestellen

Spezifikation Testergebnisse

Form Off-white Powder
Storage Store at Room Temperature (15 - 30 °C).
Source Green coffee bean or coffee seeds

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR