Daunorubicin Hydrochlorid, Pulver

Lieferant: MP Biomedicals

Synonyme: Daunorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-8-Acetyl-10-(((2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

0215077705 0215077701
ICNA0215077705EA 110 EUR
ICNA0215077705 ICNA0215077701
Daunorubicin Hydrochlorid, Pulver
Daunorubicin hydrochloride

Soluble in water, methanol, aqueous alcohols. Practically insoluble in chloroform, ether, benzene.


LD50 in mice 26 mg/kg in vitro.

Formel: C₂₇H₂₉NO₁₁·HCl
Molecular Weight: 563.98 g/mol
MDL: MFCD04974507
CAS-Nummer: 23541-50-6

Bestellen

Spezifikation Testergebnisse

Form Powder
Storage Store at +4 °C, protect from light.
Source Streptomyces peucetius


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR