5-Guanylylimidodiphosphate tetralithium salt, Pulver

Lieferant: MP Biomedicals

0215120410 0215120425 0215120405
ICNA0215120410EA 1137 EUR
ICNA0215120410 ICNA0215120425 ICNA0215120405
5-Guanylylimidodiphosphate tetralithium salt, Pulver
5-Guanylylimidodiphosphate tetralithium salt
Formel: C₁₀H₁₃Li₄N₆O₁₃P₃
CAS-Nummer: 64564-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR