4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside, Pulver

Lieferant: MP Biomedicals

Synonyme: 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside, 4-Methylumbelliferyl N-Acetyl-beta-D-glucosaminide

0215168183 0215168180
ICNA0215168183EA 300 EUR
ICNA0215168183 ICNA0215168180
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside, Pulver
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Soluble in methanol, warm water (100 mg/65 ml) and DMF (20 mg/ml).

Formel: C₁₈H₂₁NO₈
Molecular Weight: 379,365 g/mol
MDL: MFCD00065481
CAS-Nummer: 37067-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR