L-thyroxine, Pulver

Lieferant: MP Biomedicals

Synonyme: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid

02152145.1 02152145.5 0215214501 0215214505
ICNA02152145.1EA 35.3 EUR
ICNA02152145.1 ICNA02152145.5 ICNA0215214501 ICNA0215214505
L-thyroxine, Pulver
L-thyroxine

Soluble in solutions of alkali hydroxides (50 mg/ml in 4 M ammonium hydroxide in methanol); slightly insoluble in alcohol; insoluble in water, acetone, chloroform and ether.

Formel: C₁₅H₁₁I₄NO₄
Molecular Weight: 776.874 g/mol
MDL: MFCD00002595
CAS-Nummer: 51-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR