Xylazine hydrochloride, weißes Pulver
Lieferant: MP Biomedicals
|
Gefahr
|
Synonyme:
N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride, 2-[(2,6-Dimethylphenyl)amino]-5,6-dihydro-4H-1,3-thiazine Hydrochloride
Xylazine hydrochloride is an alpha agonist with sedative, analgesic and muscle relaxant properties.
- Presentation : White Powder
- Solubility : Soluble in water (50 mg/mL).
- Key Applications : To produce rapid and reversible anesthesia in experimental animals
- Product Type : Biochemicals
A solution of ketamine hydrochloride and xylazine hydrochloride has been used together to produce rapid and reversible anesthesia in experimental animals. Xylazine elicited a diuretic response in ketamine-anesthesised animals, probably through activation of complex peripheral and CNS α2-adrenergic receptor systems.
α2-Adrenergic receptor agonist, sedative, muscle relaxant.
Formel:
C₁₂H₁₆N₂S Molecular Weight: 256,79 g/mol Schmelzpunkt: 161,..171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058196 CAS-Nummer: 23076-35-9 UN: 2811 ADR: ,III |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 161 - 171°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...