Diazolidinyl urea, Pulver

Lieferant: MP Biomedicals

ICNA0219019125EA 55.7 EUR
ICNA0219019125 ICNA0219019180 ICNA0219019183
Diazolidinyl urea, Pulver
Diazolidinyl urea

Soluble in water (50% w/v).


Diazolidinyl Urea is a mixture of different formaldehyde addition products including polymers. It is chemically related to imidazolidinyl urea.


Diazolidinyl Urea is used as an antimicrobial preservative in cosmetics. It acts as a formaldehyde releaser.

Formel: C₈H₁₄N₄O₇
Molecular Weight: 278,22 g/mol
MDL: MFCD03547942
CAS-Nummer: 78491-02-8

Bestellen

Spezifikation Testergebnisse

Form White Powder
Storage Store at +4 °C
pH 6,0 - 7,5 (1% aq soln)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR