Formaldehyd 16% w/v methanol_free, Pierce™
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Methanal, Formalin, Formol, Methylaldehyd
28906 28908
PIER28906EA 60.5 EUR
PIER28906 PIER28908
Formaldehyd 16% w/v methanol_free, Pierce™
Formaldehyd
Methanol-free formaldehyde is a highly reactive, cell-permeable agent that is used by researchers as a reversible crosslinking agent for proteins and nucleic acids within the cell or as a general cell-fixing agent for imaging-based applications.
  • Formaldehyde solution remains stable and free of contamination in an amber pre-scored ampoule
  • Package sizes geared for applications ranging from culture-targeted cell fixing to in-cell protein-based applications
1 ml ampoules are intended for protein-based in cell applications such as chromatin immunoprecipitation (ChIP) or protein interaction discovery.
10 ml ampoules are intended for bioimaging (high content screening/analysis) such as fixing cells on a culture plate before image analysis.
Formel: CH₂O
Molecular Weight: 30.03 g/mol
Siedepunkt: -19.3
Schmelzpunkt: -92
Lagertemperatur: Raumtemperatur
MDL: MFCD00003274
CAS-Nummer: 50-00-0
EINECS: 200-001-8
UN: 3287
ADR: 6.1 III
Merck Index: 13,04259