Propylamine hydrochloride 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Propan-1-amine hydrochloride, 1-Aminopropane hydrochloride

221890250 221891000
ACRO221890250EA 48.5 EUR
ACRO221890250 ACRO221891000
Propylamine hydrochloride 99+%
Propylamine hydrochloride

Appearance: White to light beige Crystalline powder

Formel: C₃H₉N
Molecular Weight: 95.57 g/mol
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012911
CAS-Nummer: 556-53-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
Titration with HClO4 ≥99.0 % (On dry substance)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR