Poly(styrene sulphonic acid)sodium salt, M.W. ∼ 70.000

Lieferant: Thermo Fisher Scientific

Synonyme: Poly(sodium-p-styrenesulphonate)

222270050 222271000 222275000
ACRO222270050EA 39.7 EUR
ACRO222270050 ACRO222271000 ACRO222275000
Poly(styrene sulphonic acid)sodium salt, M.W. ∼ 70.000
Poly(styrene sulphonic acid)sodium salt

Appearance: White to tan Powder

Formel: [-CH₂CH(C₆H₄)SO₃Na)-]n
Lagertemperatur: Raumtemperatur
MDL: MFCD00084449
CAS-Nummer: 25704-18-1

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Powder
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR