5-Azacytidine 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Ladakamycin, 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one
226620500
226620010
ACRO226620500EA
45.3
EUR
ACRO226620500
ACRO226620010
5-Azacytidine 99%
5-Azacytidine
CAS No.: 320-67-2
Formel:
C₈H₁₂N₄O₅ Molecular Weight: 244,21 g/mol Schmelzpunkt: 228…230 °C (Zers.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006539 CAS-Nummer: 320-67-2 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...