Silver lactate 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Lactic acid silver salt

233330100 233330050 233330500
ACRO233330100EA 153 EUR
ACRO233330100 ACRO233330050 ACRO233330500
Silver lactate 96%
Silver lactate

CAS No.: 128-00-7

Formel: C₃H₅AgO₃
Molecular Weight: 196,938 g/mol
Schmelzpunkt: 120…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043279
CAS-Nummer: 15768-18-0

Bestellen

Spezifikation Testergebnisse

Appearance Grey-brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR