2-[2-(1-Piperazinyl)ethoxy]ethanol 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine

259110050 259110250 259111000
ACRO259110050EA 45.4 EUR
ACRO259110050 ACRO259110250 ACRO259111000
2-[2-(1-Piperazinyl)ethoxy]ethanol 95%
2-(2-(Piperazin-1-yl)ethoxy)ethanol

CAS No.: 13349-82-1

Formel: C₈H₁₈N₂O₂
Molecular Weight: 174.2407 g/mol
Siedepunkt: 150 °C (13 mmHg)
Dichte: 1.01 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00074931
CAS-Nummer: 13349-82-1

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow-brown Viscous liquid
Infrared spectrum Conforms
Titration Tertiary base ≥94 %
GC ≥94 %
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR