Hydroxychloroquin-sulfat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulphate, N4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulphate, 7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline Sulphate
263010250
263011000
ACRO263010250EA
115
EUR
ACRO263010250
ACRO263011000
Hydroxychloroquin-sulfat 98%
Hydroxychloroquine sulphate
CAS No.: 747-36-4
Formel:
C₁₈H₂₆ClN₃O Molecular Weight: 433.95 g/mol Schmelzpunkt: 240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078203 CAS-Nummer: 747-36-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (105°C, 2 h) |
Heavy metals | ≤20 ppm |
Sulfated ash | ≤0.2 % |
UV spectrophotometry | ≥97.5 % (on dried basis) |
Chloride (Cl) | ≤350 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...