4-Piperidineethanol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(2-Hydroxyethyl)piperidine, 2-(Piperidin-4-yl)ethanol
263650010
263650100
ACRO263650010EA
30.8
EUR
ACRO263650010
ACRO263650100
4-Piperidineethanol 97%
4-Piperidineethanol
Appearance: White Low melting crystalline solid
Formel:
C₇H₁₅NO Molecular Weight: 129,20 g/mol Siedepunkt: 131…136 °C (17 mmHg) Schmelzpunkt: 45…50 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006008 CAS-Nummer: 622-26-4 |
Spezifikation Testergebnisse
Appearance | White Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 45 to 50 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...