trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99+%
Lieferant: Thermo Fisher Scientific
265870010
265872500
ACRO265870010EA
332
EUR
ACRO265870010
ACRO265872500
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99+%
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
Appearance: White to off-white Crystalline powder
Formel:
C₇H₁₆ClNO Schmelzpunkt: 150…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145424 CAS-Nummer: 28250-45-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 150 to 155 °C |
Titration Argentometric | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...